Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Thermo Scientific™

Catalog No. CC30009CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30009CB 250 mg
CC30009DA 1 g
CC30009DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC30009CB Supplier Thermo Scientific Chemicals Supplier No. CC30009CB
Add to Cart

Chemical Identifiers

CAS 158985-37-6
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.379
MDL Number MFCD06411543
InChI Key PXKYXTXYWJKTSQ-UHFFFAOYSA-N
Synonym tert-butyl 4-4-hydroxymethyl phenyl piperazine-1-carboxylate, 4-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-4-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-n-boc-piperazinyl benzylalcohol, 4-4-tert-butoxycarbonyl piperazine benzyl alcohol, pubchem11318, buttpark 98\06-99, 4-1-boc-piperazin-4-yl phenylmethanol, 4-4-tert-butoxycarbonyl piperazin-1-yl benzyl alcohol
PubChem CID 2795510
IUPAC Name tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 292.38
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.