Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™

Catalog No. CC53239DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53239DA 1 g Amber Glass Bottle
CC53239CB 250 mg Amber Glass Bottle
CC53239DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC53239DA Supplier Thermo Scientific Chemicals Supplier No. CC53239DA
Add to Cart

Chemical Identifiers

CAS 864771-44-8
Molecular Formula C18H25BN2O4
Molecular Weight (g/mol) 344.22
MDL Number MFCD08437629
InChI Key MMXDKXUKSKMROG-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
PubChem CID 18525862
IUPAC Name tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 344.22
Percent Purity 97%
Chemical Name or Material tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.