Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18607306 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-306 200 L Amber Glass Bottle
18-607-298 100 mL Amber Glass Bottle
18-607-299 500 mL Amber Glass Bottle
18-607-305 2.5 L Amber Glass Bottle
18-607-301 4 L Amber Glass Bottle
18-607-303 4 x 4 L Amber Glass Bottle
18-607-304 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-306 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1188200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B1188, 75-65-0, C4H10O

Spectrum™ Chemical tert-Butyl Alcohol, Reagent , also known as 2-methyl-2-propanol, is a tertiary alcohol that is miscible with diethyl ether and ethanol. It is used as a solvent, ethanol denaturant and paint remover ingredient. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

CAS Max % 1
Boiling Point 81.9°C to 82.8°C (179.4°F to 181°F)
Packaging Amber Glass Bottle
Quantity 200 L
Residue after Evaporation 30 ppm
Percent Purity 99%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material tert-Butyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.