Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tert-Butyl Alcohol (tertiary Butanol; 2-Methyl-2-propanol), Reagents

Catalog No. 01337545
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-545 500 mL
01-337-547 1 L
01-337-546 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-545 Supplier Reagents Holdings Llc Supplier No. C2120200500A
Add to Cart
Add to Cart

Tert-Butyl Alcohol (tertiary Butanol; 2-Methyl-2-propanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym 2-Methyl-2-propanol, t-Butanol, tert-Butanol, 2-Methylpropan-2-ol
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25.3°C
Color Colorless
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.79
Boiling Point 83°C
Grade Reagent
Chemical Name or Material Tert-Butyl Alcohol (tertiary Butanol, 2-Methyl-2-propanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.