Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Dimethylphosphonoacetate 95.0+%, TCI America™
SDP

Catalog No. B509425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B509425G 25 g
B50945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B509425G Supplier TCI America Supplier No. B509425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62327-21-3
Molecular Formula C8H17O5P
Molecular Weight (g/mol) 224.19
MDL Number MFCD00042939
InChI Key SAZYDWOWLRDDRQ-UHFFFAOYSA-N
Synonym Dimethyl (Boc-methyl)phosphonate, Dimethyl (tert-Butoxycarbonylmethyl)phosphonate, Dimethylphosphonoacetic Acid tert-Butyl Ester
PubChem CID 4564495
IUPAC Name tert-butyl 2-(dimethoxyphosphoryl)acetate
SMILES COP(=O)(CC(=O)OC(C)(C)C)OC

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 g
Formula Weight 224.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Dimethylphosphonoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.