Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Glycidyl Ether 96.0+%, TCI America™
SDP

Catalog No. B1840500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1840500ML 500 mL
B184025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1840500ML Supplier TCI America Supplier No. B1840500ML

Chemical Identifiers

CAS 7665-72-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00010050
InChI Key SFJRUJUEMVAZLM-UHFFFAOYNA-N
Synonym 1-tert-Butoxy-2,3-epoxypropane
PubChem CID 24314
IUPAC Name 2-[(tert-butoxy)methyl]oxirane
SMILES CC(C)(C)OCC1CO1

Specifications

Color Colorless
Boiling Point 152°C
Quantity 500 mL
UN Number 1993
Formula Weight 130.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Glycidyl Ether
RTECSNumber RR0475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.