Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Hydroperoxide, 70% Solution in Water, Spectrum™ Chemical
SDP

Catalog No. 18607309 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-309 500 mL Amber Glass Bottle
18-607-307 25 mL Amber Glass Bottle
18-607-308 100 mL Amber Glass Bottle
18-607-310 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-309 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B2499500ML
Only null left
Add to Cart
Add to Cart

B2499, 75-91-2, C4H10O2

Spectrum™ Chemical tert-Butyl Hydroperoxide, 70 Percent Solution in Water , with a systematic name of tert-Butyl hydroperoxide, is used in a variety of oxidation processes. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 75-91-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
IUPAC Name 2-methylpropane-2-peroxol
SMILES CC(C)(C)OO

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
UN Number UN3109
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.