Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Hypochlorite 98.0+%, TCI America™
SDP

Catalog No. H0362500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0362500G 500 g
H036225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0362500G Supplier TCI America Supplier No. H0362500G

Chemical Identifiers

CAS 507-40-4
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.565
MDL Number MFCD00037105
InChI Key IXZDIALLLMRYOU-UHFFFAOYSA-N
Synonym Hypochlorous Acid tert-Butyl Ester
PubChem CID 521297
IUPAC Name tert-butyl hypochlorite
SMILES CC(C)(C)OCl

Specifications

Color Yellow
Boiling Point 78°C
Quantity 500 g
UN Number 3255
Formula Weight 108.57
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material tert-Butyl Hypochlorite
EINECSNumber (2)-2798
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.