Learn More
Sigma Aldrich Fine Chemicals Biosciences tert Butyl methyl ether HP1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 650560
Encompass
tert-Butyl methyl ether is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetics of its heat-assisted persulfate oxidation has been investigated at various pH temperature oxidant concentration and ionic strength levels. Its biodegradation followed a pseudo-first-order decay model and its pseudo-first-order rate constants were evaluated. MTBE can be synthesized by acid catalyzed reaction between methanol and isobutene. A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method.
Catalog No. 50-290-6741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.