Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-d5, for NMR, with 0.03% TMS, 100.0 atom% D

Catalog No. AC351490050
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle

Catalog No. AC351490050

Supplier: Thermo Scientific Chemicals 351490050

Only null left
Add to Cart

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7, 75-76-3
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Color Colorless
Refractive Index 1.507
Flash Point 20°C
Packaging Glass Bottle
Quantity 5 mL
Deuterium (D) 99.95% min.
Water 0.02% max. (Coulometric)
Density 1.0500g/mL
Melting Point -41.0°C
Boiling Point 114.4°C
Beilstein 20, III, 2305
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5, 100.0 atom% D
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-720-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.