Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Methyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. B147525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B147525ML 25 mL
B14755ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B147525ML Supplier TCI America Supplier No. B147525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6163-64-0
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00008840
InChI Key CJFVCTVYZFTORU-UHFFFAOYSA-N
Synonym tert-butyl methyl sulfide, propane, 2-methyl-2-methylthio, sulfide, tert-butyl methyl, methyl tert-butyl sulfide, 3,3-dimethyl-2-thiabutane, 2-methyl-2-methylsulfanyl propane, t-butyl methyl sulfide, methyl t-butyl sulfide, tert-butyl methyl sulfane, acmc-1b7tx
PubChem CID 138679
IUPAC Name 2-methyl-2-(methylsulfanyl)propane
SMILES CSC(C)(C)C

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Methyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.