Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Nitrite 90.0+%, TCI America™
SDP

Catalog No. N0357250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0357250ML 250 mL
N035725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0357250ML Supplier TCI America Supplier No. N0357250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-80-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002055
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O

Specifications

Color Yellow
Boiling Point 63°C
Quantity 250 mL
UN Number 2351
Formula Weight 103.12
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Nitrite
RTECSNumber RA0802000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.