Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Propiolate 98.0+%, TCI America™
SDP

Catalog No. P103825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103825G 25 g
P10385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P103825G Supplier TCI America Supplier No. P103825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13831-03-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00060100
InChI Key XGTPDIIFEPTULX-UHFFFAOYSA-N
Synonym tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
PubChem CID 543038
IUPAC Name tert-butyl prop-2-ynoate
SMILES CC(C)(C)OC(=O)C#C

Specifications

Color Colorless
Boiling Point 53°C
Quantity 25 g
UN Number 3272
Formula Weight 126.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Propiolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.