Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (Triphenylphosphoranylidene)carbamate 98.0+%, TCI America™
SDP

Catalog No. B568725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B568725G 25 g
B56875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B568725G Supplier TCI America Supplier No. B568725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68014-21-1
Molecular Formula C23H24NO2P
Molecular Weight (g/mol) 377.424
MDL Number MFCD00798174
InChI Key KNXPVXCUELYHDM-UHFFFAOYSA-N
Synonym (Triphenylphosphoranylidene)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)imino(triphenyl)phosphorane, N-Boc-imino(triphenyl)phosphorane
PubChem CID 2756802
IUPAC Name tert-butyl N-(triphenyl-$l^{5}-phosphanylidene)carbamate
SMILES CC(C)(C)OC(=O)N=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 149°C
Color White
Quantity 25 g
Formula Weight 377.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl (Triphenylphosphoranylidene)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.