Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylamine, 99%

Catalog No. AC107820025
Change view
Click to view available options
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107820025 2.5 L Glass Bottle
AC107820050 5 mL Glass Bottle
AC107820010 1 L Glass Bottle
AC107820100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC107820025 Supplier Thermo Scientific Chemicals Supplier No. 107820025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-64-9 | C4H11N | 73.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N

Specifications

Melting Point -67.0°C
Density 0.6900g/mL
Boiling Point 46.0°C
Flash Point −38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3770 to 1.3790
Linear Formula (CH3)3CNH2
Quantity 2.5 L
Beilstein 04,173
Fieser 01,84; 02,43; 10,62; 11,138
Merck Index 15,1547
Specific Gravity 0.69
Solubility Information Solubility in water: 1000g/L (25°C). Other solubilities: miscible with alcohol,soluble in common organic solvents
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye p
EINECSNumber 200-888-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.