Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldichlorophosphine, 98%

Catalog No. p-7029848
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2736003 1 g
AAH2736009 10 g
AAH2736018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2736003 Supplier Thermo Scientific Chemicals Supplier No. H2736003
Only null left
Add to Cart

CAS: 25979-07-1 | C4H9Cl2P | 158.99 g/mol

Reactant for the synthesis of alkali metal complexes. As intermediates. tert-Butyldichlorophosphine (TBDCP) is used primarily in the preparation of monoand bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for the synthesis of alkali metal complexes. As intermediates. tert-Butyldichlorophosphine (TBDCP) is used primarily in the preparation of monoand bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.

Solubility
Reacts with water. Soluble in n-Pentane, n-Hexane, Ether, Toluene,THF. tert-Butyldichlorophosphine is infinitely soluble in hydrocarbon and ethereal solvents.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong bases, air, water and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25979-07-1
Molecular Formula C4H9Cl2P
Molecular Weight (g/mol) 158.99
MDL Number MFCD00013615
InChI Key NMJASRUOIRRDSX-UHFFFAOYSA-N
Synonym tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
PubChem CID 117690
IUPAC Name tert-butyl(dichloro)phosphane
SMILES CC(C)(C)P(Cl)Cl

Specifications

Melting Point 44°C to 49°C
Linear Formula (CH3)3CPCl2
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water. Soluble in n-Pentane,n-Hexane,Ether,Toluene,THF. tert-Butyldichlorophosphine is infinitely soluble in hydrocarbon and ethereal solvents.
Formula Weight 159
Percent Purity 98%
Chemical Name or Material tert-Butyldichlorophosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 247-386-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.