Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilanol 95.0+%, TCI America™
SDP

Catalog No. B549925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B549925ML 25 mL
B54995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B549925ML Supplier TCI America Supplier No. B549925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18173-64-3
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00010673
InChI Key FGWRMMTYIZKYMA-UHFFFAOYSA-N
PubChem CID 554527
IUPAC Name tert-butyl-hydroxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)O

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 mL
UN Number 1987
Formula Weight 132.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyldimethylsilanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.