Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(tert-Butyldimethylsilyl)acetylene 97.0+%, TCI America™
SDP

Catalog No. B436525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B436525G 25 g
B43655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B436525G Supplier TCI America Supplier No. B436525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86318-61-8
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.301
MDL Number MFCD00191877
InChI Key RTYNRTUKJVYEIE-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
PubChem CID 2757281
IUPAC Name tert-butyl-ethynyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)C#C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
UN Number 1993
Formula Weight 140.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (tert-Butyldimethylsilyl)acetylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.