Learn More
Description
- Tert-butylsulfinyl chloride can be synthesized by using tert-butylmagnesium chloride as the starting material via a multi-step synthesis process.tert-butylsulfinyl chloride can react with primary and secondary amines to form tert-butylsulfinamides, which on oxidation forms base stable tert-butylsulfonamide
- It can also react with 1-thiosugars to form the corresponding carbohydrate thionolactones
Specifications
Specifications
| CAS | 31562-43-3 |
| Density | 1.135 g/mL (at 25°C (literature)) |
| Boiling Point | 53°C to 54°C (19 mmHg) |
| Molecular Formula | C4H9ClOS |
| Refractive Index | n20/D 1.4830 (literature) |
| Linear Formula | (CH3)3 CS(O)Cl |
| MDL Number | MFCD04974074 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 140.63 |
| Percent Purity | 97% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.