Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylsulfonamide 98.0+%, TCI America™
SDP

Catalog No. B43471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43471G Supplier TCI America Supplier No. B43471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34813-49-5
Molecular Formula C4H11NO2S
Molecular Weight (g/mol) 137.197
MDL Number MFCD02179404
InChI Key GWJSQKNYHPYZRN-UHFFFAOYSA-N
PubChem CID 2757335
IUPAC Name 2-methylpropane-2-sulfonamide
SMILES CC(C)(C)S(=O)(=O)N

Specifications

Melting Point 168°C
Color White
Quantity 1 g
Formula Weight 137.20
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylsulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.