Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Octylamine 95%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
Packaging:
Glass bottle
107-45-9
C8H19N
129.24
MFCD00008053
QIJIUJYANDSEKG-UHFFFAOYSA-N
tert-octylamine,1,1,3,3-tetramethylbutylamine,2-pentanamine, 2,4,4-trimethyl,t-octylamine,2,4,4-trimethyl-2-pentylamine,butylamine, 1,1,3,3-tetramethyl,1,1,3,3-tetramethylbutanamine,butylamine, bis 1,3-dimethyl,unii-s84lil883b,2,4,4-trimethyl-2-pentanamine
61017
2,4,4-trimethylpentan-2-amine
CC(C)(C)CC(C)(C)N
107-45-9
C8H19N
129.24
MFCD00008053
QIJIUJYANDSEKG-UHFFFAOYSA-N
tert-octylamine,1,1,3,3-tetramethylbutylamine,2-pentanamine, 2,4,4-trimethyl,t-octylamine,2,4,4-trimethyl-2-pentylamine,butylamine, 1,1,3,3-tetramethyl,1,1,3,3-tetramethylbutanamine,butylamine, bis 1,3-dimethyl,unii-s84lil883b,2,4,4-trimethyl-2-pentanamine
61017
2,4,4-trimethylpentan-2-amine
CC(C)(C)CC(C)(C)N
Water 0.3% max.
Quantity 5g
Boiling Point 137.0°C to 143.0°C
Specific Gravity 0.805
Density 0.8050g/mL
Refractive Index 1.4235 to 1.4255
Color Undesignated
Flash Point 32°C
Packaging Glass bottle
Melting Point -67.0°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula (CH3)3CCH2C(CH3)2NH2
Beilstein 04, 198
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents
Formula Weight 129.24
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material tert-Octylamine, 95%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-491-1

RTECSNumber : EO3850000

TSCA : TSCA