Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TESK1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121551
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121551 20 μL
PIPA5121550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121551 Supplier Invitrogen™ Supplier No. PA5121551
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TESK1 serine/threonine kinase or testis-specific kinase 1 contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role during and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TESK1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TESK1
Gene Accession No. O70146, Q15569, Q63572
Gene Alias AI326901; dual specificity testis-specific protein kinase 1; Tesk1; Testicular protein kinase 1; testis specific protein kinase 1; testis-specific kinase 1
Gene Symbols TESK1
Host Species Rabbit
Immunogen Phe57-Ile314 (Accession Q15569), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21754, 29460, 7016
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.