Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TESK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731562 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1562 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1562 Supplier Proteintech Group Inc Supplier No. 174141AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

TESK2 (testicular protein kinase 2) is a nuclear protein that belongs to the protein kinase superfamily and is expressed in testis and prostate. Functioning as a dual specificity protein kinase, TESK2 catalyzes the ATP-dependent phosphorylation of substrates and autophosphorylation on tyrosine and serine/ threonine residues, thereby mediating intracellular signal transduction pathways. TESK2 requires magnesium as a cofactor and its catalytic activity is thought to play an important role in meiotic events such as spermatogenesis.TESK2 contains one protein kinase domain that is 65% identical to the kinase domain found in TESK1 (testicular protein kinase 1), suggesting a similar role for these proteins in phosphorylation events. Three isoforms of TESK2 are expressed due to alternative splicing.

Specifications

Antigen TESK2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TESK2
Gene Accession No. Q8VCT9, Q924U5, Q96S53
Gene Alias TESK2, Testicular protein kinase 2, testis kinase 2
Gene Symbols Tesk2
Host Species Rabbit
Immunogen TESK2 Fusion Protein Ag10964
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10420, 170908, 230661
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.