Learn More
Med Vet International McKesson 10-Drug Panel with Adulterants Drugs of Abuse Test, 16-8105A3, 25/Box
Supplier:  Med Vet International 1101531BX
Encompass_Preferred
Drugs of Abuse Test McKesson 10-Drug Panel with Adulterants AMP, BUP, BZO, COC, mAMP/MET, MDMA, MTD, MOP300, OXY, THC (OX, pH, SG) Urine Sample 25 Tests CLIA Waived All-in-one preliminary screening test for the qualitative detection of multiple drugs and drug metabolites in urine at specific cut-off levels CLIA-waved urine drug screen Offers panel size up to 10 analytes with an additional three adulteration tests Tests simultaneously for the following drugs, consumption of which is prohibited by the Controlled Substances Act for amphetamine, benzodiazepines, buprenorphine, cocaine, marijuana, methamphetamine, ecstasy, opiates, methadone, oxycodone Rapid, visually read results in just 5 minutes Of the 10 analytes, specifically, this test cup provides screening for buprenorphine (BUP) and opiates (MOP300) at low-level cut offs Built-in specimen validity test (SVT) strip verifies the validity of the specimen to compare with the adulteration color comparison chart card (included)
Catalog No. 50-304-4718