Learn More
Eisco™ Test Strips 5-9 pH - for Weak Solutions - Quick, Accurate Narrow Range pH Testing Kit for Rainwater, Gardening, and Spa Use, Easy 1-2 Second Dip
SCI_ED
Catalog No. S02930 Shop All Eisco Products
Change view
Click to view available options
pH Range:
5 to 9
Catalog No. pH Range
S02930 5 to 9
1 options
Catalog No. S02930 Supplier Eisco™ Supplier No. CH202405
Add to Cart
Add to Cart
Quick, accurate narrow-range pH testing kit for rainwater, gardening, and spa use. Measures pH 5-9 with an easy 1-2 second dip for fast results. Ideal for measuring pH in weak solutions.
  • Precise testing: Designed for narrow range pH 5-9, these strips are perfect for testing weakly acidic to neutral solutions like rainwater and alkaline water.
  • Educational use: An excellent resource for classrooms or home science kits, demonstrating pH testing effectively with real-time results.
  • Gardening and soil check: Ideal for monitoring soil conditions in gardens. Helps ensure optimal soil pH for plant health and growth.
  • Spa and pool maintenance: Quickly verify the pH levels of your spa or pool, crucial for maintaining water quality and safety without heavy chemicals.
  • Our pH 5-9 Test Strips are tailor-made for accurately testing the pH of mildly acidic or alkaline solutions, such as rainwater, garden soil, or spa water.
  • This kit provides an efficient, reliable method for environmental monitoring, gardening enthusiasts, and pool maintenance.
  • Each test is quick and straightforward: simply dip a strip, shake off the excess, and read against the provided color chart.
  • These strips are an indispensable tool for anyone needing easy and immediate pH assessments.
  • Please note: These test strips are not intended to treat or diagnose any diseases, and are not designed for medical use.
Specifications
pH Range 5 to 9
Type pH 5-9 Test Strips for Weak Solutions