Learn More
United Scientific™ Test Tube Clamp with Finger Grips
SDPSCI_ED
Catalog No. S15716 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Brass
Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S15716 Steel
S15828 Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15716 Supplier United Scientific™ Supplier No. TTCL03
Add to Cart
Add to Cart
Test Tube Clamp for use with all standard test tube sizes.
  • Self-closing clamp is designed to securely hold test tubes.
  • They are constructed from brass wire and include finger grips for added comfort and ease of use.
Specifications
Quantity 1 Ea.
Type Test Tube Clamp
Material Steel
Color Gold
Jaw Type Spring Loaded