Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Test Tube Drying Rack
SDPSCI_ED

Catalog No. S15723 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Array:
5 x 10
8 x 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array
S15723 8 x 12
S15983 5 x 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15723 Supplier United Scientific™ Supplier No. 77851
Only null left
Add to Cart
Add to Cart

Dry your test tubes without taking up a lot of bench space.

  • These polypropylene racks can conveniently hold inverted tubes for drying and minimize collection or airborne contaminants.
  • Racks can also be used to hold and dry electrophoresis and chromatography plates.
  • Autoclavable

Specifications

Autoclavable Autoclavable
Color Red
Holds Test Tubes
Material Polypropylene
Positions 96
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 13 mm Tubes
Includes 6 Racks
Product Type Tube Rack with Drying Pins
Shelves None
Array 8 x 12
Show More Show Less

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.