Learn More
Description
- Designed for use with biologically hazardous materials
- Also applicable in research with viruses, cancerous cells and recombinant DNA
- Sterilization can be performed with tubes in racks prior to discarding hazardous materials
- Universal biohazard symbol tag attached to each rack
- Epoxy coated steel wire; chemical- and corrosion-resistant
- Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Orange |
| Holds | Tubes (13 mm) |
| Material | Epoxy-coated Steel |
| Positions | 72 |
| Shape | Rectangular |
| Stackable | No |
| Sterility | Non-sterile |
| Diameter (Metric) Opening | 13 mm |
| Product Type | Tube Rack |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.