Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab, Inc. Test Tubes

Catalog No. 22505497
Change view
Click to view available options
Capacity (Metric):
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-505-497 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-497 Supplier Marketlab, Inc. Supplier No. 105436
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quality That Measures Up. Reliable lab essentials designed for high performance in any setting.

  • Made of either premium polystyrene for added clarity or polypropylene for greater impact strength and stability when handling common acids
  • Conical bottom helps efficiently separate solid particles during centrifugation
  • Polypropylene tubes are autoclavable and freezable
  • Polystyrene (PS) has an RCF of 1500 x g
  • Polypropylene (PP) has an RCF of 3000 x g

Specifications

Capacity (Metric) 12 mL
Length (Metric) 100 mm
Material Polystyrene
Shape Conical Bottom Tube
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Product Type Test Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.