Learn More
Velp Scientifica™ Test Tubes for Digestion Units
SDP
Catalog No. 11392034
Change view
Click to view available options
For Use With (Equipment):
DK6/48 and UDK series
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
UDK Series distillation units
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Diameter (Metric) Outer
11-392-034 Digesters DK 20/26, DKL 42/26 and UDK Distillation Units 26 mm
11-392-170 DK6/48 and UDK series 48 mm
11-392-169 UDK Series distillation units 80 mm
11-392-032 - 42 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-392-034 Supplier Velp Scientifica™ Supplier No. A00000146
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
6 Test Tubes/Box
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available in different counts
  • Available in different sizes and diameters
Specifications
Product Type Test Tubes
Capacity (Metric) 100 mL
Diameter (Metric) Outer 26 mm
For Use With (Equipment) Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
Quantity 6 Tubes/ Box