Learn More
Description
- Tissue from 24 year old Male Testis in normal condition
- Store at 4°C, Do not freeze
- Slides are guaranteed for 3 months from date of receipt
- This product is for research use only and is not approved for use in humans or in clinical diagnosis
Specifications
Specifications
| Quantity | 5 |
| Specimen Type | testis tissue |
| Tissue Condition | Normal Adult |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
