Learn More
Enzo Life Sciences Testosterone ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901065
Encompass_Preferred
The Testosterone ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies. Sensitivity: 5.67 pg/ml (range 7.81 - 2,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent
Catalog No. 50-658-218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.