Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Testosterone Parameter Assay Kit
SDP

Catalog No. SKGE010 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
SKGE010 6 x 96-well Plates
KGE010 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SKGE010 Supplier R&D Systems™ Supplier No. SKGE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Testosterone Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Testosterone Parameter Assay Kit quantifies multi-species Testosterone in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).

Specifications

Assay Range 0.04 - 10ng/mL
Assay Sensitivity 0.041ng/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Testosterone Levels in Various Samples
Sufficient For 6 x 96-well Plates
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.