Learn More
Abcam TET Hydroxylase Activity Quantification Kit, Fluorometric, 96 Tests
Description
- Assay Type: Enzyme activity
- Detection Method: Fluorescent
- Assay Time: 5 hr
- Sensitivity: > 10 ng
- Range: 10 to 500 ng
- Sample Types: Nuclear Extracts, Purified protein
- Applications: Enzyme activity assay
- Cited in: > 8 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| For Use With (Equipment) | Microplate Reader |
| Target Molecule | TET1 |
| No. of Reactions | 96 Tests |
| Format | Kit |
| Detection Method | Fluorescence |
| Detection Location | In-Solution Detection |
| Sample Type | Nuclear Extracts, Purified protein |
| Quantity | 96 Tests |
| For Use With (Application) | Enzyme activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.