Learn More
Description
Recommended positive controls: Tetracyclin repressor (tetR). This antibody works in any species where the TetR system is being used to regulate gene ezpression in mammalian cells. Not suitable for immunofluorescence.
Specifications
Specifications
| Antigen | Tet Repressor |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene Alias | pSC101 tetR; TetR |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein. Sequence is 100% conserved in all TetR classes. |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
