Learn More
Clontech Labs 3P Tet System Approved FBS, US-Sourced 500 ml
Supplier: Clontech Labs 3P 631101
Fetal Bovine Serum (FBS) that has been functionally tested for optimal use with all of our Tet Systems. This FBS does not contain trace levels of tetracycline (or derivatives) which have been observed to interfere with proper regulation of TRE-controlled gene expression. Use of this serum ensures the maximal range of induction possible with the Tet Systems. This FBS has been processed in the United States. 500 ml