Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TET2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585488 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585488 Supplier Invitrogen™ Supplier No. PA585488
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, DDDDK-human TET2-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TET2 is a dioxygenase that catalyzes the conversion of the modified genomic base 5-methylcytosine (5mC) into 5-hydromethylcytosine (5hmC) and plays a key role in active DNA demethylation. TET2 has a preference for 5-hydroxymethylcytosine in CpG motifs. It mediates subsequent conversion of 5hmC into 5-formylcytosine (5fC) and conversion of 5fC to 5-carboyxlcytosine (5caC). Conversion of 5mC into 5hmC, 5fC, and 5caC probably constitutes the first step in cytonsine demethylation. In addition to TET2's role in DNA demethylation, it is also involved in the recruitment of the O-GlcNAc transferase OGT to CpG-rich transcription start sites of active genes, thereby promoting histone H2B GlcNAcylation by OGT. Mutations in the genes can result in Polycythemia vera and Myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TET2
Gene Accession No. Q4JK59, Q6N021
Gene Alias Ayu17-449; E130014J05Rik; FLJ20032; KIAA1546; MDS; methylcytosine dioxygenase TET2; MGC125715; mKIAA1546; Nbla00191; OTTHUMP00000161869; probable methylcytosine dioxygenase TET2; Protein Ayu17 449; Protein Ayu17-449; RGD1311625; Tet 2; tet methylcytosine dioxygenase 2; tet oncogene 2; tet oncogene family member 2; TET2
Gene Symbols TET2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TET2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214133, 54790
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.