Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 50560683 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-560-683 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-560-683 Supplier Proteintech Group Inc Supplier No. 212071AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes all three isoforms of TET2.

TET2 is a dioxygenase that catalyzes the conversion of the modified genomic base 5-methylcytosine (5mC) into 5-hydromethylcytosine (5hmC) and plays a key role in active DNA demethylation. TET2 has a preference for 5-hydroxymethylcytosine in CpG motifs. It mediates subsequent conversion of 5hmC into 5-formylcytosine (5fC) and conversion of 5fC to 5-carboyxlcytosine (5caC). Conversion of 5mC into 5hmC, 5fC, and 5caC probably constitutes the first step in cytonsine demethylation. In addition to TET2′s role in DNA demethylation, it is also involved in the recruitment of the O-GlcNAc transferase OGT to CpG-rich transcription start sites of active genes, thereby promoting histone H2B GlcNAcylation by OGT. Mutations in the genes can result in Polycythemia vera and Myelodysplastic syndrome.

Specifications

Antigen TET2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TET2
Gene Accession No. Q4JK59, Q6N021
Gene Alias KIAA1546, tet oncogene family member 2, TET2
Gene Symbols TET2
Host Species Rabbit
Immunogen TET2 Fusion Protein Ag15583
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214133, 54790
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.