Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetanus Toxic Fragment C Polyclonal Antibody, Rockland™
SDP

Catalog No. p-7190900
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0082 25 μL
50-105-7417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0082 Supplier Rockland Immunochemicals Supplier No. 100401894S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against the C fragment of Tetanus toxin derived from Clostridium tetani. A BLAST analysis was used to suggest cross reactivity with Clostridium tetani. Cross-reactivity with TTFC from other sources have not been determined.

Tetanus toxin C-fragment (TTc) is a non-toxic 52 kD polypeptide generated by papain cleavage of tetanus toxin, corresponding to the C-terminal 451 amino acids. The illness known as Tetanus is caused by a neurotoxin produced by the anaerobic bacterium Clostridium tetani. C. tetani is commonly found in soil, feces, and on many objects lying on the ground, such as rusty metal, hence puncture wounds are a common cause of tetanus. It acts upon the presynaptic membranes of both central and peripheral nervous systems to block the release of neurotransmitters. In its toxic form tetanus neurotoxin is a 150 kDa protein, consisting of two major components: a light chain and heavy chain. The light chain contains the enzymatic portion of the toxin and is responsible for its toxic effects. The heavy chain binds to the neuron and aids delivery of the light chain to the neuron interior. A single disulfide bond links light and heavy chains.

Specifications

Antigen Tetanus Toxic Fragment C
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 68 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a polyhistidine-tagged TTFC fusion protein expressed in E. coli corresponding to full length TTFC.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.