Learn More
Charles River Labs Cell Solutions Tetanus Toxoid (830-844) Peptide 1mg, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ANTIGEN1054
Encompass
A tetanus toxoid peptide that has been described as universally immunogenic, QYIKANSKFIGITEL is recognized by a T cell line also available from Astarte Biologics. The T cell line is found at our Antigen Specific T cells Page. The original publication in the European Journal of Immunology can be found at the publishers website.
Catalog No. 50-225-6091
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.