Learn More
Invitrogen™ Tetra Electrophoresis Primary Core
Catalog No. 1658037
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
16-580-37 1 Ea.
1 options
Catalog No. 16-580-37 Supplier Invitrogen™ Supplier No. 1658037
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart
Easily swap and run Invitrogen protein gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank with the Tetra Electrophoresis Primary Core. Benefit from high-performance separations, fast run times, and run up to four gels using a primary plus a companion core.
The Invitrogen Tetra Electrophoresis Primary Core allows Invitrogen precast protein gels to be run without the need to purchase an entirely new system or adopt a new workflow. Designed for seamless integration with Bio-Rad's Mini-PROTEAN Tetra Cell, our Tetra primary core provides an easy and straightforward method of adopting Invitrogen precast mini gels for protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.
  • Swap and go—designed to work with Bio-Rad's Mini-PROTEAN Tetra systems, allowing for a smooth transition without the need for new equipment
  • Achieve high performance—compatible with Invitrogen precast protein gels for high-quality results
  • Run multiple gels—capable of running up to four gels simultaneously to increase throughput (with a Companion Core, sold separately)
  • Access versatile gel options—compatible with all Invitrogen mini gels for diverse applications
The Tetra Electrophoresis Primary Core stands out due to its compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. Capable of running up to four gels simultaneously when combined with our Tetra Electrophoresis Companion Core, it significantly increases throughput and efficiency. Additionally, its self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.
The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel in the core. To run three or four gels at once, the Tetra Electrophoresis Companion Core (Cat. No. 1658038) is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.
This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Primary Core is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols. Invitrogen precast protein gels offer excellent protein integrity with neutral pH, a wide range of molecular weight separation, fast run times, and a long shelf life, making them ideal for high-performance and reproducible results in your experiments.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Bio-Rad Mini-PROTEAN Tetra Cell
Type Electrophoresis Core
Mode of Separation Molecular Weight
Quantity 1 Ea.
For Research Use Only.