Learn More
Tetra-n-butylammonium hydrogen sulfate, 97%
Description
Phase transfer catalyst Tetra-n-butylammonium hydrogen sulfate shows antibacterial properties due to the presence of the quaternary amine and the counter ion. It serves as a phase-transfer catalyst, surface-active agent, solvent, intermediate, active ingredient for conditioners, antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It is used as an efficient catalyst in the N-alkylation reactions of benzanilides, cyclization of beta-amino acids to beta-lactams, conversion of nitriles to amides, dehydrohalogenation of aryl 2-haloethyl ethers to give vinyl ethers and in the oxidation of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst Tetra-n-butylammonium hydrogen sulfate shows antibacterial properties due to the presence of the quaternary amine and the counter ion. It serves as a phase-transfer catalyst, surface-active agent, solvent, intermediate, active ingredient for conditioners, antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It is used as an efficient catalyst in the N-alkylation reactions of benzanilides, cyclization of beta-amino acids to beta-lactams, conversion of nitriles to amides, dehydrohalogenation of aryl 2-haloethyl ethers to give vinyl ethers and in the oxidation of alcohols.
Solubility
Soluble in water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 168°C to 172°C |
| Quantity | 1000 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 339.54 |
| Percent Purity | 97% |
| Chemical Name or Material | Tetra-n-butylammonium hydrogen sulfate |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 251-068-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.