Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraammineplatinum(II) hydroxide solution, Pt 8-11% w/w (cont. Pt)

Catalog No. AA42918NB
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42918NB 5 g
AA42918NH 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42918NB Supplier Thermo Scientific Chemicals Supplier No. 042918NB
Only null left
Add to Cart

CAS: 38201-97-7 | H14N4O2Pt | 297.22 g/mol

Tetraammineplatinum(II) hydroxide solution acts as a precursor for a platinum catalyst. It acts as noble metal catalyst which is primarily useful for homogeneous catalysis applications. It is used as a catalyst in hydrogenation, carbonylation, hydroformylation, chiral catalysis reactions and ruthenium dyes. Furthermore, it is used for C-X coupling reaction like Heck, Suzuki and Stille reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38201-97-7
Molecular Formula H14N4O2Pt
Molecular Weight (g/mol) 297.22
MDL Number MFCD00050044
InChI Key VSKCDODNDPOZKS-UHFFFAOYSA-L
IUPAC Name platinum(2+) tetraamine dihydroxide
SMILES N.N.N.N.[OH-].[OH-].[Pt++]

Specifications

Color Colorless
Odor Odorless
Linear Formula (NH3)4Pt(OH)2
Quantity 5 g
Solubility Information Miscible with water.
Formula Weight 297.23
Concentration or Composition (by Analyte or Components) Pt 8 to 11% w/w (cont. Pt)
Physical Form Liquid
Chemical Name or Material Tetraammineplatinum (II) hydroxide solution
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P362-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 253-823-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.