Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabenazine 99.0+%, TCI America™
SDP

Catalog No. T28391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28391G 1 g
T2839200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28391G Supplier TCI America Supplier No. T28391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-46-8
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.429
InChI Key MKJIEFSOBYUXJB-UHFFFAOYSA-N
Synonym 3-Isobutyl-9,10-dimethoxy-1,3,4,6,7,11b-hexahydro-2H-benzo[a]quinolizin-2-one
PubChem CID 6018
ChEBI CHEBI:64028
IUPAC Name 9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b-hexahydrobenzo[a]quinolizin-2-one
SMILES CC(C)CC1CN2CCC3=CC(=C(C=C3C2CC1=O)OC)OC

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 1 g
Formula Weight 317.43
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabenazine
EINECSNumber (9)-2283
RTECSNumber DK2275000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.