Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromo-o-sulfobenzoic Anhydride 98.0+%, TCI America™
SDP

Catalog No. T004825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T004825G 25 g
T00485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T004825G Supplier TCI America Supplier No. T004825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68460-01-5
Molecular Formula C7Br4O4S
Molecular Weight (g/mol) 499.749
MDL Number MFCD00009745
InChI Key QPGYGIVRJIICGZ-UHFFFAOYSA-N
PubChem CID 110335
IUPAC Name 4,5,6,7-tetrabromo-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)S(=O)(=O)OC2=O

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 499.75
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromo-o-sulfobenzoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.