Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium chloride hydrate, 96%

Catalog No. AC432060250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432060250 25 g Glass bottle
AC432060050 5 g Glass bottle
AC432061000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC432060250 Supplier Thermo Scientific Chemicals Supplier No. 432060250
Add to Cart
This item is not returnable. View return policy

Phase transfer catalyst | CAS: 37451-68-6 | C16H36ClN | 277.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37451-68-6
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
PubChem CID 16212273
IUPAC Name tetrabutylazanium;chloride;hydrate
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 37.0°C to 39.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (On dried substance) (Argentometry)
Packaging Glass bottle
Quantity 25 g
Formula Weight 295.94
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material Tetrabutylammonium chloride hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.