Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Difluorotriphenylsilicate 97.0+%, TCI America™
SDP

Catalog No. T190925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T190925G 25 g
T19095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T190925G Supplier TCI America Supplier No. T190925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 163931-61-1
Molecular Formula C34H51F2NSi
Molecular Weight (g/mol) 539.871
MDL Number MFCD00274218
InChI Key RQBKGJOQACIQDG-UHFFFAOYSA-N
Synonym TBAT
PubChem CID 9893474
IUPAC Name difluoro(triphenyl)silanuide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Si-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F

Specifications

Melting Point 156°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 539.87
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Difluorotriphenylsilicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.