Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Dihydrogen Trifluoride 90.0+%, TCI America™
SDP

Catalog No. T163525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T163525G 25 g
T16355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T163525G Supplier TCI America Supplier No. T163525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
IUPAC Name tetrabutylazanium dihydrofluoride fluoride
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Yellow
Quantity 25 g
UN Number 1760
Formula Weight 301.48
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Dihydrogen Trifluoride
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.