Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Diiodoaurate, TCI America™
SDP

Catalog No. T13571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13571G 1 g
T1357100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13571G Supplier TCI America Supplier No. T13571G

Chemical Identifiers

CAS 50481-03-3
Molecular Formula C16H36AuI2N
Molecular Weight (g/mol) 693.25
MDL Number MFCD00145387
InChI Key IPICULSEVGYZBD-UHFFFAOYSA-L
Synonym tetrabutylammonium diiodoaurate, tetra-n-butylammonium diiodoaurate, tetrabutylammonium diiodoaurate i
PubChem CID 44630513

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 693.25
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Diiodoaurate
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.