Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydrogen Sulfate 98.0+%, TCI America™
SDP

Catalog No. I0368100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0368100G 100 g
I036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0368100G Supplier TCI America Supplier No. I0368100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name tetrabutylazanium hydrogen sulfate
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 173°C
Color White
Quantity 100 g
Formula Weight 339.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Hydrogen Sulfate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-0186
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.